5-Bromo-2-chloro-1H-1,3-benzimidazole ≥85%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-chloro-1H-benzo[d]imidazole

042893-1G
101858-084EA 60 USD
101858-084
5-Bromo-2-chloro-1H-1,3-benzimidazole ≥85%
5-Bromo-2-chloro-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₇H₄BrClN₂
MW: 231.49 g/mol
Melting Pt: 212…213 °C
MDL Number: MFCD04128987
CAS Number: 683240-76-8

Order Now

Specification Test Results

Melting point 212-213°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR