6-Amino-2-(4-bromophenyl)imidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

042892-500MG
101858-078EA 1109.23 USD
101858-078
6-Amino-2-(4-bromophenyl)imidazo[1,2-a]pyridine ≥95%
6-Amino-2-(4-bromophenyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₁₃H₁₀BrN₃
Melting Pt: 191…195 °C
MDL Number: MFCD08276939
CAS Number: 885950-52-7

Order Now

Specification Test Results

Melting point 191-195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR