3-(3-Bromophenyl)-5-methyl-1H-pyrazole ≥95%

Supplier: Matrix Scientific

042891-1G
101858-074EA 461.54 USD
101858-074
3-(3-Bromophenyl)-5-methyl-1H-pyrazole ≥95%
3-(3-Bromophenyl)-5-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₁₀H₉BrN₂
Melting Pt: 101…104 °C
MDL Number: MFCD08276940
CAS Number: 324054-75-3

Order Now

Specification Test Results

Melting point 101-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR