Methyl-4-bromo-1-methyl-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-bromo-1-methyl-1H-pyrrole-2-carboxylate

042890-5G
101858-070EA 429.23 USD
101858-070
Methyl-4-bromo-1-methyl-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-bromo-1-methyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₇H₈BrNO₂
MW: 218.06 g/mol
Melting Pt: 63…65 °C
MDL Number: MFCD08275077
CAS Number: 1196-90-3

Order Now

Specification Test Results

Melting point 63-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR