2-Amino-4'-bromoacetophenone hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromophenacylamine hydrochloride

042886-1G
101858-046EA 350.77 USD
101858-046
2-Amino-4'-bromoacetophenone hydrochloride ≥95%
2-Amino-4'-bromoacetophenone hydrochloride

Hazard: Irritant

Formula: C₈H₉BrClNO
MW: 250.53
Melting Pt: 274…276 °C
MDL Number: MFCD00051998
CAS Number: 5467-72-1

Order Now

Specification Test Results

Melting point 274-276°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR