Ethyl-2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(4-methoxyphenyl)thiazole-4-carboxylate

042885-5G 042885-500MG
101858-038EA 375.38 USD
101858-038 101858-042
Ethyl-2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate ≥95%
Ethyl 2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₃NO₃S
MW: 263.3100 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD06738343
CAS Number: 57677-79-9

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR