2-Chloro-5-iodonicotinic acid ≥95%

Supplier: Matrix Scientific

042883-1G
101858-030EA 461.54 USD
101858-030
2-Chloro-5-iodonicotinic acid ≥95%
2-Chloro-5-iodonicotinic acid

Hazard: Irritant

Formula: C₆H₃ClINO₂
MW: 283.4500 g/mol
Melting Pt: 221…224 °C
MDL Number: MFCD08276959
CAS Number: 59782-86-4

Order Now

Specification Test Results

Melting point 221-224°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR