2'-Formylbiphenyl-4-carboxylic acid ethyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2'-formyl-[1,1'-biphenyl]-4-carboxylate
042881-1G
101858-018EA
467.69
USD
101858-018
2'-Formylbiphenyl-4-carboxylic acid ethyl ester ≥95%
2'-Formylbiphenyl-4-carboxylic acid ethyl ester
Hazard: Irritant
Formula:
C₁₆H₁₄O₃ Melting Pt: 45…48 °C |
MDL Number:
MFCD08276944 CAS Number: 885950-48-1 |
Specification Test Results
Melting point | 45-48°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...