3'-Nitro[1,1'-biphenyl]-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(3-Nitrophenyl)benzaldehyde
042876-1G
101857-994EA
350.77
USD
101857-994
3'-Nitro[1,1'-biphenyl]-4-carbaldehyde ≥95%
3'-Nitro[1,1'-biphenyl]-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₃H₉NO₃ MW: 227.22 g/mol Melting Pt: 112…114 °C |
MDL Number:
MFCD06803015 CAS Number: 411206-92-3 |
Specification Test Results
Melting point | 112-114°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...