4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbohydrazide ≥95%

Supplier: Matrix Scientific

042874-1G
101857-982EA 698.46 USD
101857-982
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbohydrazide ≥95%
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbohydrazide

Hazard: Irritant

Formula: C₁₂H₁₀F₃N₃OS
Melting Pt: 170…173 °C
MDL Number: MFCD00112085
CAS Number: 263387-25-3

Order Now

Specification Test Results

Melting point 170-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR