4-(4-Nitrophenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine ≥95%
Supplier: Matrix Scientific
042871-1G
101857-970EA
350.77
USD
101857-970
4-(4-Nitrophenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine ≥95%
4-(4-Nitrophenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₂S Melting Pt: 108…110 °C |
MDL Number:
MFCD06797378 CAS Number: 213462-01-2 |
Specification Test Results
Melting point | 108-110°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...