2-Butyl-1,3-benzoxazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

042870-1G
101857-966EA 402.5 USD
101857-966
2-Butyl-1,3-benzoxazole-5-carboxylic acid ≥95%
2-Butyl-1,3-benzoxazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
Melting Pt: 136…138 °C
MDL Number: MFCD06797377
CAS Number: 885949-50-8

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR