3-(Dimethylamino)-2-(4-methylphenyl)acrylaldehyde ≥95%

Supplier: Matrix Scientific

042868-1G
101857-954EA 232.31 USD
101857-954
3-(Dimethylamino)-2-(4-methylphenyl)acrylaldehyde ≥95%
3-(Dimethylamino)-2-(4-methylphenyl)acrylaldehyde

Hazard: Irritant

Formula: C₁₂H₁₅NO
Melting Pt: 133…135 °C
MDL Number: MFCD07366535
CAS Number: 53868-38-5

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR