N-[2-(Aminocarbonyl)phenyl]-4-chloro-3-nitrobenzenecarboxamide ≥95%
Supplier: Matrix Scientific
Synonyms:
N-[2-(Aminocarbonyl)phenyl]-4-chloro-3-nitrobenzamide
042865-5G
042865-500MG
101857-934EA
375.38
USD
101857-934
101857-938
N-[2-(Aminocarbonyl)phenyl]-4-chloro-3-nitrobenzenecarboxamide ≥95%
N-[2-(Aminocarbonyl)phenyl]-4-chloro-3-nitrobenzenecarboxamide
Hazard: Irritant
Formula:
C₁₄H₁₀ClN₃O₄ MW: 319.71 g/mol Melting Pt: 271…272 °C |
MDL Number:
MFCD00495266 CAS Number: 331240-40-5 |
Specification Test Results
Melting point | 271-272°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...