4-Ethoxy-2-phenyl-5-pyrimidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

042863-500MG 042863-5G
101857-922EA 353.85 USD
101857-922 101857-926
4-Ethoxy-2-phenyl-5-pyrimidinecarboxylic acid ≥95%
4-Ethoxy-2-phenyl-5-pyrimidinecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₃
Melting Pt: 181…183 °C
MDL Number: MFCD06797376
CAS Number: 136326-10-8

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR