6-(4-Chlorophenyl)-3-morpholinone ≥95%

Supplier: Matrix Scientific

042860-500MG
101857-906EA 921.54 USD
101857-906
6-(4-Chlorophenyl)-3-morpholinone ≥95%
6-(4-Chlorophenyl)-3-morpholinone

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₂
MW: 211.65 g/mol
Melting Pt: 142…145 °C
MDL Number: MFCD06797375
CAS Number: 5196-95-2

Order Now

Specification Test Results

Melting point 142-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR