3-Amino-4-chloro-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-1H-indazol-3-amine

042849-1G
101857-850EA 83.08 USD
101857-850
3-Amino-4-chloro-1H-indazole ≥95%
3-Amino-4-chloro-1H-indazole

Hazard: Irritant

Formula: C₇H₆ClN₃
MW: 167.60 g/mol
Melting Pt: 159…162 °C
MDL Number: MFCD00663139
CAS Number: 20925-60-4

Order Now

Specification Test Results

Melting point 159-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR