tert-Butyl-4-(4-aminobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-butyl 4-(4-aminobenzoyl)piperazine-1-carboxylate

042846-1G
101857-838EA 615.38 USD
101857-838
tert-Butyl-4-(4-aminobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(4-aminobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₃N₃O₃
MW: 305.38 g/mol
Melting Pt: 159…160 °C
MDL Number: MFCD06659531
CAS Number: 350684-49-0

Order Now

Specification Test Results

Melting point 159-160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR