Ethyl-2-(5-bromo-1H-indol-3-yl)-2-oxoacetate ≥95%
Supplier: Matrix Scientific
042842-5G
042842-500MG
101857-814EA
123.08
USD
101857-814
101857-818
Ethyl-2-(5-bromo-1H-indol-3-yl)-2-oxoacetate ≥95%
Ethyl-2-(5-bromo-1H-indol-3-yl)-2-oxoacetate
Hazard: Irritant
Formula:
C₁₂H₁₀BrNO₃ MW: 296.13 g/mol Melting Pt: 248…249 °C |
MDL Number:
MFCD02657946 CAS Number: 17826-11-8 |
Specification Test Results
Melting point | 248-249°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...