Methyl-2-amino-5-(4-chlorophenyl)-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

042841-1G
101857-810EA 189.23 USD
101857-810
Methyl-2-amino-5-(4-chlorophenyl)-1,3-thiazole-4-carboxylate ≥95%
Methyl-2-amino-5-(4-chlorophenyl)-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₂S
MW: 268.72 g/mol
Melting Pt: 238…240 °C
MDL Number: MFCD06200914
CAS Number: 127918-92-7

Order Now

Specification Test Results

Melting point 238-240°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR