5-(Chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole ≥95%

Supplier: Matrix Scientific

042812-1G 042812-5G 042812-500MG
101857-668EA 556.92 USD
101857-668 101857-672 101857-670
5-(Chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole ≥95%
5-(Chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₉H₆Cl₂N₂O
Melting Pt: 61…63 °C
MDL Number: MFCD00119077
CAS Number: 57238-75-2

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR