Pentafluorophenyl 4-iodobenzenesulfonate ≥95%

Supplier: Matrix Scientific

042810-1G 042810-500MG 042810-5G
101857-658EA 349.6 USD
101857-658 101857-656 101857-660
Pentafluorophenyl 4-iodobenzenesulfonate ≥95%
Pentafluorophenyl 4-iodobenzenesulphonate

Hazard: Irritant

Formula: C₁₂H₄F₅IO₃S
Melting Pt: 88…90 °C
MDL Number: MFCD05975132
CAS Number: 885950-39-0

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR