Pentafluorophenyl 3-methylbenzenesulfonate ≥95%

Supplier: Matrix Scientific

042802-5G 042802-500MG 042802-1G
101857-612EA 1073.85 USD
101857-612 101857-610 101857-608
Pentafluorophenyl 3-methylbenzenesulfonate ≥95%
Pentafluorophenyl 3-methylbenzenesulphonate

Hazard: Irritant

Formula: C₁₃H₇F₅O₃S
Melting Pt: 49…51 °C
MDL Number: MFCD05975125
CAS Number: 885950-34-5

Order Now

Specification Test Results

Melting point 49-51°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR