Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentafluorophenyl 2-thiophenesulfonate ≥95%

Supplier: Matrix Scientific


042801-1G 042801-500MG 042801-5G
101857-604EA 397.54 USD InStock
101857-604 101857-602 101857-606
Pentafluorophenyl 2-thiophenesulfonate ≥95%
Pentafluorophenyl 2-thiophenesulphonate
Purity: 95%
CAS: 663175-91-5
MDL Number: MFCD05975124
Molecular Formula: C10H3F5O3S2
Molecular Weight: 330,25
Hazard: Irritant


Formula: C₁₀H₃F₅O₃S₂
Melting Pt: 90…92 °C
MDL Number: MFCD05975124
CAS Number: 663175-91-5
EINECS: 000-000-0
Order Now
Specification Test Results
Melting point 90-92°C


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty