Pentafluorophenyl 4-chlorobenzenesulfonate ≥95%

Supplier: Matrix Scientific

042800-1G 042800-500MG 042800-5G
101857-600EA 802.7 USD
101857-600 101857-598 101857-596
Pentafluorophenyl 4-chlorobenzenesulfonate ≥95%
Pentafluorophenyl 4-chlorobenzenesulphonate

Hazard: Irritant

Formula: C₁₂H₄ClF₅O₃S
Melting Pt: 56…58 °C
MDL Number: MFCD05975123
CAS Number: 161759-99-5

Order Now

Specification Test Results

Melting point 56-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR