6-(Ethylsulfonyl)-2-(2-hydroxyethyl)-2H-1,4-benzoxazin-3(4H)-one ≥95%

Supplier: Matrix Scientific

042796-500MG 042796-5G 042796-1G
101857-576EA 532.31 USD
101857-576 101857-574 101857-572
6-(Ethylsulfonyl)-2-(2-hydroxyethyl)-2H-1,4-benzoxazin-3(4H)-one ≥95%
6-(Ethylsulfonyl)-2-(2-hydroxyethyl)-2H-1,4-benzoxazin-3(4H)-one

Hazard: Irritant

Formula: C₁₂H₁₅NO₅S
Melting Pt: 145…147 °C
MDL Number: MFCD05974884
CAS Number: 860648-57-3

Order Now

Specification Test Results

Melting point 145-147°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR