3-[2-(2-Cyanophenoxy)phenyl]acrylic acid ≥95%
Supplier: Matrix Scientific
042786-1G
042786-5G
042786-500MG
101857-528EA
927.69
USD
101857-528
101857-526
101857-530
3-[2-(2-Cyanophenoxy)phenyl]acrylic acid ≥95%
3-[2-(2-Cyanophenoxy)phenyl]acrylic acid
Hazard: Irritant
Formula:
C₁₆H₁₁NO₃ Melting Pt: 214…216 °C |
MDL Number:
MFCD03844846 CAS Number: 851975-09-2 |
Specification Test Results
Melting point | 214-216°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...