tert-Butyl 3-[(methylsulfonyl)oxy]-1-azetanecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-[(Methylsulphonyl)oxy]azetidine-1-carboxylate1-(tert-Butoxycarbonyl)-3-(mesyloxy)azetidinetert-butyl 3-(methanesulphonyloxy)azetidine-1-carboxylatetert-butyl 3-(methanesulfonyloxy)azetidine-1-carboxylate3-[(Methylsulfonyl)oxy]azetidine-1-carboxylic acid tert-Butyl estertert-Butyl 3-[(Methylsulfonyl)oxy]-1-azetanecarboxylate3-[(Methylsulphonyl)oxy]azetidine-1-carboxylic acid tert-Butyl estertert-Butyl 3-[(Methylsulfonyl)oxy]azetidine-1-carboxylatetert-Butyl 3-[(Methylsulphonyl)oxy]-1-azetanecarboxylate1-Boc-3-(mesyloxy)azetidine

042782-1G
101857-506EA 89.7 USD
101857-506
tert-Butyl 3-[(methylsulfonyl)oxy]-1-azetanecarboxylate ≥95%
tert-Butyl 3-[(methylsulfonyl)oxy]-1-azetanecarboxylate

Hazard: Irritant

Formula: C₉H₁₇NO₅S
MW: 251.3 g/mol
Melting Pt: 65…67 °C
MDL Number: MFCD05664832
CAS Number: 141699-58-3

Order Now

Specification Test Results

Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR