4-(2-Fluorobenzoyl)-1-methyl-1H-pyrrole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
042766-1G
042766-5G
042766-500MG
101857-418EA
278.46
USD
101857-418
101857-422
101857-420
4-(2-Fluorobenzoyl)-1-methyl-1H-pyrrole-2-carbaldehyde ≥95%
4-(2-Fluorobenzoyl)-1-methyl-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₀FNO₂ Melting Pt: 122…123 °C |
MDL Number:
MFCD03425711 CAS Number: 446275-97-4 |
Specification Test Results
Melting point | 122-123°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...