6-(1-Naphthyl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

042753-5G 042753-500MG 042753-1G
101857-354EA 532.31 USD
101857-354 101857-352 101857-350
6-(1-Naphthyl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid ≥95%
6-(1-Naphthyl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₂N₂O₃
Melting Pt: 248…250 °C
MDL Number: MFCD03425708
CAS Number: 439094-93-6

Order Now

Specification Test Results

Melting point 248-250°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR