Ethyl-2,4-bis(trifluoromethyl)imidazo-[1,2-a][1,8]naphthyridine-8-carboxylate ≥95%
Supplier: Matrix Scientific
042751-5G
042751-500MG
042751-1G
101857-338EA
375.38
USD
101857-338
101857-342
101857-340
Ethyl-2,4-bis(trifluoromethyl)imidazo-[1,2-a][1,8]naphthyridine-8-carboxylate ≥95%
Ethyl-2,4-bis(trifluoromethyl)imidazo-[1,2-a][1,8]naphthyridine-8-carboxylate
Hazard: Irritant
Formula:
C₁₅H₁₁F₆N₃O₂ MW: 379.26 g/mol Melting Pt: 189…191 °C |
MDL Number:
MFCD28385260 CAS Number: 439094-91-4 |
Specification Test Results
Melting point | 189-191°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...