3-(3-Methoxyphenyl)-1-oxo-3,4-dihydro-1H-isochromene-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

042747-5G 042747-500MG 042747-1G
101857-318EA 377.2 USD
101857-318 101857-316 101857-314
3-(3-Methoxyphenyl)-1-oxo-3,4-dihydro-1H-isochromene-4-carboxylic acid ≥95%
3-(3-Methoxyphenyl)-1-oxo-3,4-dihydro-1H-isochromene-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₄O₅
Melting Pt: 114…116 °C
MDL Number: MFCD03001219
CAS Number: 400751-55-5

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR