N-(2-Cyanophenyl)thiourea ≥97%
Supplier: Matrix Scientific
042718-1G
042718-500MG
101857-170EA
345
USD
101857-170
101857-168
N-(2-Cyanophenyl)thiourea ≥97%
N-(2-Cyanophenyl)thiourea
Hazard: Irritant
Formula:
C₈H₇N₃S Melting Pt: 281…284 °C |
MDL Number:
MFCD03791138 CAS Number: 851950-53-3 |
Specification Test Results
Melting point | 281-284°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...