3-Chloro-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-5-(trifluoromethyl)-2(1H)-pyridinone ≥95%
Supplier: Matrix Scientific
Synonyms:
5,5'-Bis(trifluoromethyl)-3,3'-dichloro-2H-1,2'-bipyridin-2-one
042713-500MG
042713-5G
042713-1G
101857-146EA
1196
USD
101857-146
101857-144
101857-142
3-Chloro-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-5-(trifluoromethyl)-2(1H)-pyridinone ≥95%
3-Chloro-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-5-(trifluoromethyl)-2(1H)-pyridinone
Hazard: Irritant
Formula:
C₁₂H₄Cl₂F₆N₂O MW: 377.07 g/mol Melting Pt: 107…109 °C |
MDL Number:
MFCD01568202 CAS Number: 96741-18-3 |
Specification Test Results
Melting point | 107-109°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...