4-Chloro-N-hydroxybenzenecarboximidoyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: alpha,4-Dichlorobenzaldoxime

042691-500MG 042691-5G 042691-1G
101857-034EA 602.6 USD
101857-034 101857-032 101857-030
4-Chloro-N-hydroxybenzenecarboximidoyl chloride ≥95%
4-Chloro-N-hydroxybenzenecarboximidoyl chloride

Hazard: Irritant

Formula: C₇H₅Cl₂NO
MW: 190.03 g/mol
Melting Pt: 87…88 °C
MDL Number: MFCD00136374
CAS Number: 28123-63-9

Order Now

Specification Test Results

Melting point 87-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR