4-Chloro-2-(trifluoromethyl)quinazoline ≥95%

Supplier: Matrix Scientific

042685-1G
101857-000EA 423.08 USD
101857-000
4-Chloro-2-(trifluoromethyl)quinazoline ≥95%
4-Chloro-2-(trifluoromethyl)quinazoline

Hazard: Irritant

Formula: C₉H₄ClF₃N₂
MW: 232.5900 g/mol
Melting Pt: 65…68 °C
MDL Number: MFCD00793683
CAS Number: 52353-35-2

Order Now

Specification Test Results

Melting point 65-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR