Dimethyl-2-(2-nitrophenyl)malonate ≥95%
Supplier: Matrix Scientific
042673-1G
042673-500MG
042673-5G
101856-928EA
181.54
USD
101856-928
101856-930
101856-932
Dimethyl-2-(2-nitrophenyl)malonate ≥95%
Dimethyl-2-(2-nitrophenyl)malonate
Hazard: Irritant
Formula:
C₁₁H₁₁NO₆ MW: 253.21 g/mol Melting Pt: 48…50 °C |
MDL Number:
MFCD03617780 CAS Number: 26465-37-2 |
Specification Test Results
Melting point | 48-50°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...