2-Amino-3-cyano-4,5-di(furyl)furan ≥95%
Supplier: Matrix Scientific
Synonyms:
5'-Amino-2,2':3',2''-terfuran-4'-carbonitrile
042672-1G
042672-500MG
042672-5G
101856-922EA
181.54
USD
101856-922
101856-924
101856-926
2-Amino-3-cyano-4,5-di(furyl)furan ≥95%
2-Amino-3-cyano-4,5-di(furyl)furan
Hazard: Irritant
Formula:
C₁₃H₈N₂O₃ MW: 240.22 g/mol Melting Pt: 183…185 °C |
MDL Number:
MFCD00099027 CAS Number: 24386-17-2 |
Specification Test Results
Melting point | 183-185°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...