3-Chloro-5-(trifluoromethyl)-2-pyridinylhydrosulfide ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-2-thio-5-(trifluoromethyl)pyridine

042664-5G 042664-1G 042664-500MG
101856-882EA 278.46 USD
101856-882 101856-884 101856-886
3-Chloro-5-(trifluoromethyl)-2-pyridinylhydrosulfide ≥95%
3-Chloro-5-(trifluoromethyl)-2-pyridinylhydrosulfide

Hazard: Irritant

Formula: C₆H₃ClF₃NS
MW: 213.61 g/mol
Melting Pt: 156…158 °C
MDL Number: MFCD00153104
CAS Number: 76041-74-2

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR