Methyl-2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate monohydrate ≥95%

Supplier: Matrix Scientific

042662-1G 042662-500MG 042662-5G
101856-870EA 278.46 USD
101856-870 101856-872 101856-874
Methyl-2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate monohydrate ≥95%
Methyl-2-amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate monohydrate

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₃S
Melting Pt: 105…108 °C
MDL Number: MFCD03701479
CAS Number: 449778-52-3

Order Now

Specification Test Results

Melting point 105-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR