Ethyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

042660-500MG 042660-5G 042660-1G
101856-860EA 349.6 USD
101856-860 101856-862 101856-858
Ethyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylate ≥95%
Ethyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₀F₃NO₂S
MW: 301.29 g/mol
Melting Pt: 93…96 °C
MDL Number: MFCD00142038
CAS Number: 132089-39-5

Order Now

Specification Test Results

Melting point 93-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR