2,4-Bis(trifluoromethyl)imidazo[1,2-a][1,8]naphthyridine-8-carboxylic acid ≥95%

Supplier: Matrix Scientific

042655-5G 042655-1G 042655-500MG
101856-830EA 375.38 USD
101856-830 101856-832 101856-834
2,4-Bis(trifluoromethyl)imidazo[1,2-a][1,8]naphthyridine-8-carboxylic acid ≥95%
2,4-Bis(trifluoromethyl)imidazo[1,2-a][1,8]naphthyridine-8-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₅F₆N₃O₂
Melting Pt: 281…283 °C
MDL Number: MFCD03425718
CAS Number: 439094-96-9

Order Now

Specification Test Results

Melting point 281-283°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR