2-Styrylbenzenol ≥95%

Supplier: Matrix Scientific

042650-1G 042650-500MG
101856-806EA 391 USD
101856-806 101856-804
2-Styrylbenzenol ≥95%
2-Styrylbenzenol

Hazard: Irritant

Formula: C₁₄H₁₂O
Melting Pt: 144…146 °C
MDL Number: MFCD00155163
CAS Number: 18493-15-7

Order Now

Specification Test Results

Melting point 144-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR