Methyl-4-amino-5-bromo-2-methoxybenzenecarboxylate ≥95%
Supplier: Matrix Scientific
042646-1G
042646-500MG
042646-5G
101856-782EA
453.85
USD
101856-782
101856-784
101856-786
Methyl-4-amino-5-bromo-2-methoxybenzenecarboxylate ≥95%
Methyl-4-amino-5-bromo-2-methoxybenzenecarboxylate
Hazard: Irritant
Formula:
C₉H₁₀BrNO₃ Melting Pt: 152…154 °C |
MDL Number:
MFCD02082951 CAS Number: 111049-68-4 |
Specification Test Results
Melting point | 152-154°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...