2,4-Dioxo-1-phenylhexahydro-5-pyrimidinecarboximidamide ≥95%

Supplier: Matrix Scientific

042636-1G 042636-5G 042636-500MG
101856-730EA 3212.31 USD
101856-730 101856-726 101856-728
2,4-Dioxo-1-phenylhexahydro-5-pyrimidinecarboximidamide ≥95%
2,4-Dioxo-1-phenylhexahydro-5-pyrimidinecarboximidamide

Hazard: Irritant

Formula: C₁₁H₁₂N₄O₂
Melting Pt: 246…248 °C
MDL Number: MFCD00172914
CAS Number: 446276-08-0

Order Now

Specification Test Results

Melting point 246-248°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR