2,4-Dioxo-1-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%
Supplier: Matrix Scientific
042635-5G
042635-1G
042635-500MG
101856-720EA
558.9
USD
101856-720
101856-722
101856-724
2,4-Dioxo-1-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%
2,4-Dioxo-1-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile
Hazard: Irritant
Formula:
C₁₁H₇N₃O₂ Melting Pt: 296…299 °C |
MDL Number:
MFCD00172909 CAS Number: 6275-84-9 |
Specification Test Results
Melting point | 296-299°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...