1-(4-Chlorophenyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%
Supplier: Matrix Scientific
042634-1G
042634-5G
042634-500MG
101856-718EA
1591.6
USD
101856-718
101856-714
101856-716
1-(4-Chlorophenyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%
1-(4-Chlorophenyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile
Hazard: Irritant
Formula:
C₁₁H₆ClN₃O₂ Melting Pt: 255…258 °C |
MDL Number:
MFCD00172907 CAS Number: 75837-75-1 |
Specification Test Results
Melting point | 255-258°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...