1-(2-Methyl-3-nitrophenyl)-1H-pyrrole ≥95%

Supplier: Matrix Scientific

042631-500MG 042631-5G 042631-1G
101856-696EA 181.54 USD
101856-696 101856-698 101856-700
1-(2-Methyl-3-nitrophenyl)-1H-pyrrole ≥95%
1-(2-Methyl-3-nitrophenyl)-1H-pyrrole

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
Melting Pt: 70…72 °C
MDL Number: MFCD02665235
CAS Number: 52414-57-0

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR