2-(2-Chlorophenyl)-1-hydrazinecarbothioamide ≥95%
Supplier: Matrix Scientific
042627-1G
042627-500MG
042627-5G
101856-680EA
532.31
USD
101856-680
101856-676
101856-678
2-(2-Chlorophenyl)-1-hydrazinecarbothioamide ≥95%
2-(2-Chlorophenyl)-1-hydrazinecarbothioamide
Hazard: Irritant
Formula:
C₇H₈ClN₃S Melting Pt: 183…186 °C |
MDL Number:
MFCD03791140 CAS Number: 877-52-1 |
Specification Test Results
Melting point | 183-186°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...