4-(4-Fluorobenzoyl)-1-methyl-1H-pyrrole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
042589-5G
042589-500MG
101856-494EA
800
USD
101856-494
101856-490
4-(4-Fluorobenzoyl)-1-methyl-1H-pyrrole-2-carbaldehyde ≥95%
4-(4-Fluorobenzoyl)-1-methyl-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₀FNO₂ Melting Pt: 120…122 °C |
MDL Number:
MFCD00172167 CAS Number: 128843-61-8 |
Specification Test Results
Melting point | 120-122°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...